missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-ICOS/CD278 PE-Cy7™, Clone: ISA-3, Invitrogen™
Descripción
ICOS/CD278 Monoclonal antibody A14783 from Invitrogen specifically detects ICOS/CD278 in Human samples. It is validated for Flow Cytometry.
CD278 belongs to the CD28 and CTLA-4 cell-surface receptor family and is highly expressed in activated T-cells. It is essential both for efficient interaction between T and B-cells and for normal antibody responses to T-cell dependent antigens. It enhances all basic T-cell responses to a foreign antigen, namely proliferation, secretion of lymphokines, up-regulation of molecules that mediate cell-cell interaction, and effective help for antibody secretion by B-cells.
Host Species: Mouse
Cross Reactivity: Human
Isotype: IgG1
Clone: ISA-3
Immunogen: Human ICOS
Formulation: 25 Tests Purified Antibody in proprietary buffer, with 0.09% sodium azide
Synonyms: AILIM, CD278, CVID1, activation-inducible lymphocyte immunomediatory molecule, inducible T-cell costimulator, inducible costimulator
Uniprot Accession: Q9Y6W8
Entrez GeneID: 29851
Gene Symbol: ICOS
This product detects ICOS/CD278 in Human samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is Human ICOS.
Host Species: Mouse
Cross Reactivity: Human
Isotype: IgG1
Clone: ISA-3
Immunogen: Human ICOS
Formulation: 25 Tests Purified Antibody in proprietary buffer, with 0.09% sodium azide
Synonyms: AILIM, CD278, CVID1, activation-inducible lymphocyte immunomediatory molecule, inducible T-cell costimulator, inducible costimulator
Uniprot Accession: Q9Y6W8
Entrez GeneID: 29851
Gene Symbol: ICOS
This product detects ICOS/CD278 in Human samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is Human ICOS.
Especificaciones
Especificaciones
| Antígeno | ICOS/CD278 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | ISA-3 |
| Conjugado | PE-Cyanine7 |
| Formulación | 25 Tests Purified Antibody in proprietary buffer, with 0.09% sodium azide |
| N.º de referencia del gen | Q9Y6W8 |
| Alias de gen | AILIM, CD278, CVID1, activation-inducible lymphocyte immunomediatory molecule, inducible T-cell costimulator, inducible costimulator |
| Símbolos de los genes | ICOS |
| Especie del huésped | Mouse |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?