Learn More
IgG3 (Hinge region) Mouse anti-Human, HRP, Invitrogen™
Código de producto. 15235979
Change view
Click to view available options
Cantidad:
1 mL
Tamaño de la unidad:
1 ml
15235979 1 mL 1 ml
1 options
Código de producto. 15235979
Marca: Invitrogen MA516718
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
Mouse Monoclonal Secondary Antibody
Thermo Scientific Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Human IgG3 (Hinge region)
Aplicaciones ELISA, Immunohistochemistry (Frozen), Western Blot
Clasificación Monoclonal
Clon HP6050
Conjugado HRP
Especie del huésped Mouse
Inmunógeno purified Human IgG3 myeloma proteins
Método de purificación Purified
Cantidad 1 mL
Estado normativo RUO
Primario o secundario Secondary
Especies diana Human
Contenido y almacenamiento 4°C or -20°C if preferred
Formulario Liquid
Isotype IgG1