Learn More
anti-HSPC152, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Marca:  Abnova H00051504-PW2.1Set
Código de producto. 16133414
  • 843.00€ / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-HSPC152 (300μL), Antibody pair for WB: mouse purified polyclonal anti-HSPC152 (50μg)
Especificaciones
HSPC152
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
HSPC152
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-HSPC152 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-HSPC152 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Immunoprecipitation, Western Blot
Unconjugated
HSPC152
Mouse, Rabbit
RUO
51504
Human
Liquid
Certificados
Promociones
Promociones
For Research Use Only