Learn More
HPV-16 L1 Mouse anti-Human, Unlabeled, Clone: CAMVIR-1, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 550840
Descripción
More than 60 different types of human papilloma viruses (HPVs) have been isolated. The human papilloma viruses encode three late proteins, which are produced only in terminally differentiating keratinocytes, two of which (the L1 and L2 proteins) are struc
Host Species: Mouse
Clone: CamVir-1
Isotype: IgG2a κ
Species Reactivity: Human
Immunohistochemistry, Immunohistochemistry (Frozen)
Especificaciones
| HPV-16 L1 | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| 1 mL | |
| Cell Biology | |
| Human | |
| IgG2a κ |
| Immunohistochemistry | |
| CAMVIR-1 | |
| Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
For Research Use Only.