Learn More
homeobox A11, Mouse, Clone: 7E12, Abnova™
Descripción
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. This gene is involved in the regulation of uterine development and is required for female fertility. Mutations in this gene can cause radio-ulnar synostosis with amegakaryocytic thrombocytopenia. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | homeobox A11 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Clon | 7E12 |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against a partial recombinant HOXA11. |
| Formulación | PBS with no preservative; pH 7.4 |
| génica | HOXA11 |
| N.º de referencia del gen | NM_005523 |
| Alias de gen | HOX1/HOX1I |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.