Learn More
HLA-DR Mouse anti-Human, PerCP-Cy5.5, Clone: L243 (G46-6), BD
Descripción
Clone G46-6 cross-reacts with the human class II antigen of the major histocompatability complex (MHC). This clone also cross-reacts with a subset of peripheral blood lymphocytes and monocytes, but not granulocytes, of baboon and both rhesus and cynomolgus macaque monkeys. The distribution on lymphocytes is similar to that observed with human peripheral blood lymphocytes, with the majority of CD20-positive cells and a small subset of CD3-positive cells being positive with G46-6. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G46-6
Isotype: IgG2a κ
Species Reactivity: Human
Flow Cytometry
Especificaciones
Especificaciones
| Antígeno | HLA-DR |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | L243 (G46-6) |
| Conjugado | PerCP-Cyanine5.5 |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Método de purificación | Affinity Purified |
| Cantidad | 50 Tests |
| Estado normativo | RUO |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.