missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Hepatitis B Virus Core Antigen Monoclonal Antibody (1-5)

Código de producto. 11535312
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11535312 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11535312 Proveedor Invitrogen™ N.º de proveedor MA17607

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

MA1-7607 detects HBcAg from virus samples. MA1-7607 has been successfully used in Western blot, ELISA, immunoprecipitation and immunohistochemistry procedures. The MA1-7607 immunogen is a purified recombinant HBcAg core protein. The epitope corresponds to residues surrounding residue 80. Store at 4°C short term, and -80°C long term.

The hepatitis B virus is an important cause of acute and chronic liver disease. It is also the most common form of parenterally transmitted viral hepatitis. Hepatitis B virus is spherical in shape with a diameter of 42nm. It contains a 27 nm partially double stranded DNA core enclosed within a lipoprotein coat. The antigenic activity of the nucleocapsid core is designated as hepatitis B core antigen. The antigens in the outer surface are called as hepatitis B virus surface antigens. Core antigens are localized within the nuclei whereas the surface antigens are present in the cytoplasm of the infected cells. Antibodies to surface antigens appear in circulation at an early stage of infection whereas the antibodies to the core antigens are detected after several weeks. The virus is estimated to affect some 350 million people worldwide. Immunization against hepatitis B is an important component of preventative medicine and is offered to all children in approximately 30 countries. Hepatitis B virus (HBV) belongs to the Hepadnaviridae family and infection with HBV is usually more serious than infection with hepatitis A virus. In some individuals the infection is not cleared and followed by persistent intracellular viral carriage that can lead to the development of hepatocelular carcinomas. There are currently 400 million HBV carriers worldwide making HBV one of the most common human pathogens.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Hepatitis B Virus Core Antigen
Aplicaciones ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Clasificación Monoclonal
Clon 1-5
Concentración 1.0 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative; pH 7.2
Alias de gen ayw; HBc; HBcAg; HBV; HepB
Especie del huésped Mouse
Inmunógeno Purified recombinant HBcAg.
Método de purificación Protein A
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Virus
Contenido y almacenamiento 4°C short term, -80°C long term
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2b
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.