Learn More
glycine cleavage system protein H (aminomethyl carrier), Mouse, Clone: 3D8-A12, Abnova™
Descripción
The enzyme system for cleavage of glycine (glycine cleavage system; EC 2.1.2.10), which is confined to the mitochondria, is composed of 4 protein components: P protein (a pyridoxal phosphate-dependent glycine decarboxylase; MIM 238300), H protein (a lipoic acid-containing protein), T protein (a tetrahydrofolate-requiring enzyme; MIM 238310), and L protein (a lipoamide dehydrogenase; MIM 238331). Glycine encephalopathy (GCE; MIM 605899), also called nonketotic hyperglycinemia (NKH), may be due to a defect in any one of these enzymes.[supplied by OMIM
Especificaciones
Especificaciones
| Antígeno | glycine cleavage system protein H (aminomethyl carrier) |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Clon | 3D8-A12 |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against a full length recombinant GCSH. |
| Formulación | PBS with no preservative; pH 7.4 |
| génica | GCSH |
| N.º de referencia del gen | BC000790 |
| Alias de gen | GCE/NKH |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.