Learn More
GATA binding protein 2, Mouse, Clone: 1G5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GATA2.
Marca: Abnova H00002624-M02.100ug
Descripción
This gene encodes a member of the GATA family of zinc-finger transcription factors that are named for the consensus nucleotide sequence they bind in the promoter regions of target genes. The encoded protein plays an essential role in regulating transcription of genes involved in the development and proliferation of hematopoietic and endocrine cell lineages. Alternative splicing results in multiple transcript variants
Sequence: MEVAPEQPRWMAHPAVLNAQHPDSHHPGLAHNYMEPAQLLPPDEVDVFFNHLDSQGNPYYANPAHARARVSYSPAHARLTGGQMCRPHLLHSPGLPWLDGGKEspecificaciones
| GATA binding protein 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC018988 | |
| GATA2 | |
| GATA2 (AAH18988, 1 a.a. ∼ 102 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 1G5 | |
| Mouse monoclonal antibody raised against a partial recombinant GATA2. | |
| GATA2 | |
| MGC2306/NFE1B | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 2624 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.