Learn More
FOXO1A, Mouse, Clone: 1F11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FOXO1A.
Marca: Abnova H00002308-M02.100ug
Descripción
This gene belongs to the forkhead family of transcription factors which are characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in myogenic growth and differentiation. Translocation of this gene with PAX3 has been associated with alveolar rhabdomyosarcoma. [provided by RefSeq
Sequence: TQVKTPVQVPLPHPMQMSALGGYSSVSSCNGYGRMGLLHQEKLPSDLDGMFIERLDCDMESIIRNDLMDGDTLDFNFDNVLPNQSFPHSVKTTTHSWVSGEspecificaciones
| FOXO1A | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_002015 | |
| FOXO1 | |
| FOXO1A (NP_002006, 556 a.a. ∼ 655 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunofluorescence, Western Blot | |
| 1F11 | |
| Mouse monoclonal antibody raised against a partial recombinant FOXO1A. | |
| FOXO1 | |
| FKH1/FKHR/FOXO1A | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2308 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.