Learn More
anti-FMC63, scFv Antibody, Clone: Y45, Precision Avi, ACROBiosystems
Código de producto. 17256210
Change view
Click to view available options
Cantidad:
200 Tests
25 Tests
Tamaño de la unidad:
200 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17256210 25 Tests 25 tests
17246210 200 Tests 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17256210 Proveedor ACROBiosystems N.º de proveedor FM3BY5425tests
HEK293 Monoclonal Antibody
Specifically recognizes the antigen-recognition domain of FMC63 derived CARs.
Especificaciones
Antígeno FMC63
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon Y45
Conjugado Biotin
Formulación Lyophilized from 0.22μm filtered solution in PBS, 0.5% BSA, pH7.4. Normally trehalose is added as protectant before lyophilization.
Especie del huésped Human (HEK293)
Cantidad 25 Tests
Estado normativo RUO
Primario o secundario Primary
Especificidad de la prueba Specifically recognizes the antigen-recognition domain of FMC63 derived CARs.
Contenido y almacenamiento -20°C
Tipo de producto Anti-idiotypic Antibodies
Formulario Powder
Isotype IgG1 κ