Learn More
FEV, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human FEV protein.
Marca: Abnova H00054738-B02P.50ug
Descripción
This gene belongs to the ETS transcription factor family. ETS family members have a highly conserved 85-amino acid ETS domain that binds purine-rich DNA sequences. The alanine-rich C-terminus of this gene indicates that it may act as a transcription repressor. This gene is exclusively expressed in neurons of the central serotonin (5-HT) system, a system implicated in the pathogeny of such psychiatric diseases as depression, anxiety, and eating disorders. In some types of Ewing tumors, this gene is fused to the Ewing sarcoma (EWS) gene following chromosome translocations. [provided by RefSeq
Sequence: MRQSGASQPLLINMYLPDPVGDGLFKDGKNPSWGPLSPAVQKGSGQIQLWQFLLELLADRANAGCIAWEGGHGEFKLTDPDEVARRWGERKSKPNMNYDKLSRALRYYYDKNIMSKVHGKRYAYRFDFQGLAQACQPPPAHAHAAAAAAAAAAAAQDGALYKLPAGLAPLPFPGLSKLNLMAASAGVAPAGFSYWPGPGPAATAAAATAALYPSPSLQPPPGPFGAVAAASHLGGHYHEspecificaciones
| FEV | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human FEV protein. | |
| FEV | |
| HSRNAFEV/PET-1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 54738 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_017521.2 | |
| FEV | |
| FEV (NP_059991.1, 1 a.a. ∼ 238 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.