Learn More
FAK Mouse, Unlabeled, Clone: 77/FAK, BD
Descripción
Focal Adhesion Kinase (FAK) is a cytoplasmic tyrosine kinase that colocalizes with integrins in focal adhesions. This cellular localization is directed by a 125 amino acid sequence at the C-terminus called the “Focal Adhesion Targeting sequence” (FAT). The binding of extracellular matrix ligands to integrins triggers autophosphorylation and activation of FAK. This creates binding sites for SH2 domains of intracellular signaling molecules such as src, PI3 kinase, and Grb2. FAK's ability to bind numerous structural and signaling proteins via a variety of interactions has led to substantial speculation about its function. Although FAK's precise role has not been elucidated, proposed possibilities include regulating cell motility, cell growth, cytoskeletal organization, and adhesion-dependent cell survival.
Host Species: Mouse
Clone: 77
Isotype: IgG1
Species Reactivity: Human
Immunogen: Chicken FAK aa. 354-533
Formula Weight [Chemical]: 116-125kDa
Bioimaging, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Especificaciones
Especificaciones
| Antígeno | FAK |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 77/FAK |
| Concentración | 250μg/mL |
| Conjugado | Unconjugated |
| Descripción | Focal Adhesion Kinase |
| Formulación | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Chicken FAK aa. 354-533 |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.