Learn More
coagulation factor VIII, procoagulant component, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Descripción
This gene encodes coagulation factor VIII, which participates in the intrinsic pathway of blood coagulation; factor VIII is a cofactor for factor IXa which, in the presence of Ca+2 and phospholipids, converts factor X to the activated form Xa. This gene produces two alternatively spliced transcripts. Transcript variant 1 encodes a large glycoprotein, isoform a, which circulates in plasma and associates with von Willebrand factor in a noncovalent complex. This protein undergoes multiple cleavage events. Transcript variant 2 encodes a putative small protein, isoform b, which consists primarily of the phospholipid binding domain of factor VIIIc. This binding domain is essential for coagulant activity. Defects in this gene results in hemophilia A, a common recessive X-linked coagulation disorder. [provided by RefSeq]
Especificaciones
Especificaciones
| Antígeno | coagulation factor VIII, procoagulant component |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Rabbit polyclonal antibody raised against a full-length human F8 protein. |
| Formulación | PBS with no preservative; pH 7.4 |
| génica | F8 |
| N.º de referencia del gen | NM_019863.2 |
| Alias de gen | AHF/DXS1253E/F8B/F8C/FVIII/HEMA |
| Símbolos de los genes | F8 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.