Learn More
DEAD (Asp-Glu-Ala-Asp) box polypeptide 54 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant DDX54.
Marca: Abnova H00079039-A01.50uL
Descripción
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The nucleolar protein encoded by this gene interacts in a hormone-dependent manner with nuclear receptors, and represses their transcriptional activity. Alternative splice variants that encode different isoforms have been found for this gene. [provided by RefSeq
Sequence: DDRDSDEEGASDRRGPERRGGKRDRGQGASRPHAPGTPAGRVRPELKTKQQILKQRRRAQKLHFLQRGGLKQLSARNRRRVQELQQGAFGRGARSKKGKMRKRMEspecificaciones
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 54 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant DDX54. | |
| DDX54 | |
| DP97/MGC2835 | |
| Mouse | |
| 50 μL | |
| Signal Transduction | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_024072 | |
| DDX54 | |
| DDX54 (NP_076977, 778 a.a. ∼ 881 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 79039 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.