missing translation for 'onlineSavingsMsg'
Learn More

Cytochrome c Mouse, Unlabeled, Clone: 6H2.B4, BD

Código de producto. 15808698
Change view
Click to view available options
Cantidad:
0.1 mg
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15808698 0.1 mg cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15808698

Marca: BD Biosciences 556432

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

A cytochrome is an electron-transporting protein that contains a heme prosthetic group. Cytochromes have been known to be essential components of the mitochondrial respiratory chain since 1925. The iron atom of the heme group in cytochromes alternates between a reduced ferrous (+2) state and an oxidized ferric (+3) state during electron transport in oxidative phosphorylation. Cytochromes are classified into four groups (a, b, c and d) according to spectrochemical characteristics, and there are five cytochromes between coenzyme QH2 and O2 in the electron transport chain. Cytochrome c is a water-soluble protein that either promotes cell survival or death, depending upon its intracellular location. In healthy cells, it is a peripheral membrane protein of the mitochondria that transports electrons from the coenzyme QH2 cytochrome c reductase complex to the cytochrome c oxidase complex. When proapoptotic stimuli induce breakdown of the mitochondria, cytochrome c is released to the cytosol where it functions in the activation of caspases that trigger apoptosis. The 6H2.B4 monoclonal antibody has been reported to recognize the native and not the denatured form of rat, mouse, and human cytochrome c. Furthermore, studies utilizing competitive ELISA indicate that mAb 6H2.B4 binds to a region around residue 62 of rat cytochrome c. The 6H2.B4 monoclonal antibody is not useful for western blot analysis. For the western blot application, clone 7H8.2C12 (Mfr.no. 556433) would be recommended. Suggested positive controls for detecting cytochrome c include P388D mouse lymphoma cells (ATCC CCL 46),
HeLa human carcinoma cells (ATCC CCL2), Jurkat T leukemia cells (ATCC TIB152) and NIH 3T3 mouse fibroblast cells (ATCC CRL 1658). This antibody is routinely tested by immunoprecipitation and immunofluorescent imaging. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 6H2.B4
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Rat cytochrome c

Immunoprecipitation

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Cytochrome c
Aplicaciones Immunoprecipitation
Clasificación Monoclonal
Clon 6H2.B4
Concentración 0.5mg/mL
Conjugado Unconjugated
Formulación Aqueous buffered solution containing ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Rat cytochrome c
Método de purificación Affinity Purified
Cantidad 0.1 mg
Estado normativo RUO
Disciplina de investigación Cell Biology
Primario o secundario Primary
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store undiluted at 4°C
Isotype IgG1 κ
Mostrar más Mostrar menos

For Research Use Only.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.