missing translation for 'onlineSavingsMsg'
Learn More

Cyclin C Rabbit, Unlabeled, Polyclonal, BD

Código de producto. 15809648
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15809648 0.1 mL cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15809648

Marca: BD Biosciences 558903

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Cyclins and cyclin-dependent kinases (cdks) are evolutionarily conserved proteins that are essential for cell-cycle control in eukaryotes. Cyclins contain a conserved amino acid sequence motif, the cyclin box, which allows their binding to cdks to form active complexes that regulate progression of the cell cycle. More recently, cyclins are being shown to have additional functions not restricted to cell cycle regulation. Thus, cyclins have been placed into functional groups as follows: Group 1 cyclins (A, B, D1, D2, D3, E and F) function primarily in cell cycle reguation; Group 2 cyclins (C and H) also play a role in transcriptional regulation; Group 3 cyclins (G1, G2 and I) may play a role distinct from either cell cycle or transcriptional regulation. Cyclin C has been identified in both human and chicken species. Like many cyclins, the expression of cyclin C oscillates throughout the cell cycle, with a peak observed during G1 phase. Cyclin C may play a dual role within the cell in its ability to regulate both cell cycle progression as well as gene transcription. Cyclin C associates with Cdk8, forming a complex which can induce gene transcription of Cdc2 (Cdk1). Because Cdc2 kinase is important for cell entry into mitosis, cyclin C's ability to regulate cell cycle progression may be attributed, in part, to modulation of Cdc2 protein expression.6 Cyclin C has a predicted molecular weight of 36kDa. The polyclonal antiserum reacts with human and mouse cyclin C. A polypeptide fragment containing amino acids 290-303 (QGPNGSQNSSYSQS) at the C-terminus of human cyclin C, with the addition of a C-terminal cysteine (C) residue to facilitate conjugation to KLH, was used as immunogen.

Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antígeno Cyclin C
Aplicaciones Western Blot
Clasificación Polyclonal
Clon POLYCLONAL
Conjugado Unconjugated
Formulación Aqueous buffered solution containing ≤0.09% sodium azide.
Especie del huésped Rabbit
Inmunógeno Human Cyclin C polypeptide fragment aa. 290-303
Método de purificación Affinity Purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cell Biology
Primario o secundario Primary
Especies diana Human, Mouse
Contenido y almacenamiento Store undiluted at 4°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.