missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTLA-4 Mouse anti-Human, Clone: 48815, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Marca: R&D Systems MAB325-100
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CTLA-4 Monoclonal specifically detects CTLA-4 in Human samples. It is validated for Western Blot.Especificaciones
| CTLA-4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD, CD152, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA4, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1493 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 48815 | |
| Western Blot 1 ug/mL | |
| Q6GR94 | |
| CTLA4 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CTLA-4 Ala37-Phe162 (Thr174Ala) Accession # Q6GR94 | |
| 100 μg | |
| Primary | |
| Detects human CTLA-4 in direct ELISAs and Western blots. Shows approximately 10% cross-reactivity with recombinant mouse CTLA-4 and no cross-reactivity with recombinant human (rh) CD28 or rhICOS. | |
| Human | |
| Supernatant |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
For Research Use Only