missing translation for 'onlineSavingsMsg'
Learn More

COX-2 Mouse anti-Human, PE, Clone: AS67, BD

Código de producto. 15874099
Change view
Click to view available options
Cantidad:
50 Tests
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15874099 50 Tests cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15874099 Proveedor BD Biosciences N.º de proveedor 565125

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The AS67 monoclonal antibody specifically binds to Cyclooxygenase-2 (COX-2/COX2) which is also known as, Prostaglandin G/H synthase 2 (PGHS-2). The COX-2 enzyme is encoded by the PTGS2 gene, Prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase and cyclooxygenase) and exists as a homodimer that is located on the lumenal surface of the endoplasmic reticulum and on the inner and outer membranes of the nuclear envelope. COX-2 catalyzes the conversion of arachidonic acid into prostaglandin H2 (PGH2). Specific prostaglandin synthases then convert the PGH2 intermediate into various biologically active prostanoids, including Prostaglandin E2 (PGE2) and Thromboxane A2 (TXA2). COX-2 expression is not detected in most tissues under physiological conditions . However, COX-2 expression is rapidly upregulated during the course of inflammation, following cellular stresses, and in response to growth factors, tumor promoters, hormones, bacterial endotoxins, and inflammatory cytokines, eg, Interleukin-1α (IL-1α). C OX-2 can be induced in a number of cell types, including fibroblasts, endothelial cells, monocytes, ovarian follicles and mesenchymal stromal cells. Although COX-2 is structurally homologous to Cyclooxygenase-1 (COX-1), the AS67 antibody does not crossreact with recombinant human COX-1 protein as tested by immunoassay.

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno COX-2
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon AS67
Conjugado PE
Descripción hCox-2; COX2; PTGS2; PGG/HS; PGHS-2; PHS-2; GRIPGHS
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Especie del huésped Mouse
Inmunógeno Human COX-2 Recombinant Protein
Método de purificación Affinity Purified
Cantidad 50 Tests
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.