Learn More
ceramide kinase (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant CERK.
Marca: Abnova H00064781-A01.50uL
Descripción
CERK converts ceramide to ceramide 1-phosphate (C1P), a sphingolipid metabolite. Both CERK and C1P have been implicated in various cellular processes, including proliferation, apoptosis, phagocytosis, and inflammation (Kim et al., 2006 [PubMed 16488390]).[supplied by OMIM
Sequence: MQRGPASGDPREGPPRQRREGCWGLHTCLITAARLRQSGWEHDLGVCSGSTDDKLWKLAVCKALASLLLLKCQIPMLYIDAKCLTQPGLGAVRRKLAIRGGGAGPGLPQDSSEAGPEVCTRGPDLSLSFLHTEFSIFIEHLVLLSQSSVNCVSDVCLLPRSHDGSLVSSAARGLRRPEDSSRKAFLPRSPGHPSIVYYVLVEspecificaciones
| ceramide kinase | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant CERK. | |
| CERK | |
| DKFZp434E0211/FLJ21430/FLJ23239/KIAA1646/LK4/MGC131878/dA59H18.2/dA59H18.3/hCERK | |
| Mouse | |
| 50 μL | |
| Endocrine/Metabolism | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC004278 | |
| CERK | |
| CERK (AAH04278, 1 a.a. ∼ 201 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 64781 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.