Learn More
Cdk1/Cdc2 (pY15) Mouse, Unlabeled, Clone: 44, BD
Mouse Monoclonal Antibody
321.35€
Especificaciones
| Antígeno | Cdk1/Cdc2 (pY15) |
|---|---|
| Clon | 44 |
| Concentración | 250μg/mL |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|
15886159
|
BD Biosciences
612306 |
50 μg |
322.00€ 321.35€ / cada uno Ahorro 0.65€ 0.20% Descuento |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
Progression of the mammalian cell cycle is regulated by phosphorylation of many key proteins. Several classes of cyclins (A-E) act as regulatory subunits for cyclin-dependent kinases (cdks). Cdk1/Cdc2 (p34 [cdc2]) is the catalytic subunit of the maturation promoting factor (MPF), which includes the regulatory subunit cyclin B. During late S and G2 phase, cyclin B synthesis increases, allowing it to bind Cdc2. This begins the transition into M-phase of the mammalian cell cycle by initiating a series of phosphorylation and dephosphorylation events that lead to activation of the Cdc2/cyclin B complex. After binding to cyclin B, cdc2 is phosphorylated on Thr-14, by Myt1, and Tyr-15, by wee1 or mik1, yielding an inactive pre-MPF complex during G2 phase. Phosphorylation of cdc2 on Thr-161 is performed by a cdk7/cyclin H complex and is necessary for activation of the cdc2 complex. Dephosphorylation of Thr-14 and Tyr-15 by CDC25 occurs at the end of G2 phase and completes activation of the cdc2/Cyclin B complex and facilitates entry into mitosis. During mitosis, cyclin B is targeted for degradation and Cdc2 becomes inactive again.
Immunofluorescence, Western Blotting
Especificaciones
| Cdk1/Cdc2 (pY15) | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified | |
| p34 [cdc2] |
| 44 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Phosphorylated Human Cdc2 (pY15) Peptide | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.