Learn More
CD96 Mouse anti-Human, BB515, Clone: 6F9, BD
Descripción
The 6F9 monoclonal antibody specifically binds to human CD96, also known as TACTILE (T cell activation increased late expression). CD96 is a type I transmembrane glycoprotein and member of the Ig superfamily. CD96 is expressed at low levels on resting natural killer (NK) cells and T cells and at high levels on activated NK and T cells. CD96 is also expressed on some T-cell leukemia and acute myeloid leukemia cells. CD96 may serve as a marker for acute myelogenous leukemia stem cells. CD96 plays a role in the adhesive interactions of activated NK and T cells during immune responses. CD96 binds to the poliovirus receptor (CD155) that is highly expressed by some tumor cells. CD155-mediated ligation of CD96 can induce NK cell-mediated cytotoxicity. CD96-mediated uptake of CD155 may adversely affect NK cells and thus reduce their effectiveness in anti-tumor responses.
The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (eg, 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.
Especificaciones
Especificaciones
| Antígeno | CD96 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 6F9 |
| Conjugado | Brilliant Blue 515 (BB515) |
| Descripción | TACTILE; T cell activation increased late expression |
| Formulación | Aqueous buffered solution containing ≤0.09% sodium azide |
| Especie del huésped | Mouse |
| Inmunógeno | Human CD96 Transfected Cell Line |
| Método de purificación | Affinity Purified |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.