missing translation for 'onlineSavingsMsg'
Learn More

CD9 Mouse anti-Human, Unlabeled, Clone: M-L13, BD

Código de producto. 15848108
Change view
Click to view available options
Cantidad:
0.1 mg
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15848108 0.1 mg cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15848108

Marca: BD Biosciences 555370

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Reacts with a 24kDa type III transmembrane protein which is expressed on platelets, pre-B cells, monocytes, endothelia, and epithelia. CD9 belongs to a family of membrane proteins called tetraspanins which transverse the membrane four times. CD9 is weakly expressed on resting mature B cells. M-L13 induces platelet aggregation and activation. This antibody is also suitable for staining acetone-fixed, frozen tissue sections.

Host Species: Mouse
Clone: M-L13
Isotype: IgG1 κ
Species Reactivity: Hu

Flow Cytometry, Immunohistochemistry (Frozen), Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antígeno CD9
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon M-L13
Concentración 0.5mg/mL
Conjugado Unconjugated
Formulación Aqueous buffered solution containing ≤0.09% sodium azide.
Especie del huésped Mouse
Método de purificación Affinity Purified
Cantidad 0.1 mg
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.