missing translation for 'onlineSavingsMsg'
Learn More

anti-CD8 Qdot™ 605, Clone: 3B5, Invitrogen™

CD8 Monoclonal antibody Q10481 from Invitrogen specifically detects CD8 in Human samples. It is validated for Flow Cytometry.

Marca:  Invitrogen Q10481

Código de producto. 11360792

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

CD8 Monoclonal antibody Q10481 from Invitrogen specifically detects CD8 in Human samples. It is validated for Flow Cytometry.

CD8 molecule is composed of two chains termed alpha and beta. CD8 is found on a T cell subset of normal cytotoxic/suppressor cells which make up approximately 20 to 35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer cells, 80% of thymocytes, on a subpopulation of 30% of peripheral blood null cells and 15 to 30% of bone marrow cells. CD38 (T10) is a single chain 46 kDa type II integral glycoprotein with a short N terminal cytoplasmic tail. CD38 is highly expressed on thymocytes. It is also expressed by early cells of B and T lineages, NK cells, plasma cells, monocytes and macrophages and may be detected on cells from multiple myeloma, ALL (B and T) and some AML. In normal lymph nodes and tonsils the antigen is detected mainly on B cells in germinal centers and in plasma cells. The extracellular domain of the molecule shares a high homology sequence with Aplysia ADP ribosyl cyclase. CD38 functions as a multicatalytic ectoenzyme serving as ADP ribosyl cyclase, ADPR hydrolase and possibly NAD+ glycohydrolase, or as a surface receptor.
Host Species: Mouse
Cross Reactivity: Human
Isotype: IgG2a
Clone: 3B5
Immunogen: Recognizes the CD8 antigen
Formulation: 25μL Purified Antibody in 0.05M borate, with 1M betaine and 0.05% sodium azide; pH8.3
Synonyms: CD8, Leu-2, MAL, p32, CD8a, CD8b, CD8alpha, CD8beta
Uniprot Accession: P01732, P10966
Entrez GeneID: 925, 926
Gene Symbol: CD8A
This product detects CD8 in Human samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is Recognizes the CD8 antigen.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

CD8
Monoclonal
Qdot 605
P01732, P10966
CD8A
Recognizes the CD8 antigen
Primary
Human
Purified
Flow Cytometry
3B5
25μL Purified Antibody in 0.05M borate, with 1M betaine and 0.05% sodium azide; pH8.3
CD8, Leu-2, MAL, p32, CD8a, CD8b, CD8alpha, CD8beta
Mouse
25 μL
925, 926
Store at 4°C, do not freeze
IgG2a
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Promociones

Promociones

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado