missing translation for 'onlineSavingsMsg'
Learn More

CD8 Mouse anti-Human, APC-H7, Clone: SK1, BD

Código de producto. 12673356
Change view
Click to view available options
Cantidad:
100 Tests
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
12673356 100 Tests cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 12673356

Brand: BD Biosciences 641400

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD8 clone SK1 is derived from hybridization of mouse NS-1 myeloma cells with spleen cells from BALB/c mice immunized with human peripheral blood T lymphocytes.

The CD8 antibody recognizes the 32-kilodalton (kDa) α-subunit of a disulfide-linked bimolecular complex. The majority of peripheral blood CD8+ T lymphocytes express an α/β heterodimer (Mr 32, 30 kDa), while CD8

The CD8 antibody recognizes the 32-kilodalton (kDa) α-subunit of a disulfide-linked bimolecular complex. The majority of peripheral blood CD8+ T lymphocytes express an α/β heterodimer (Mr 32, 30 kDa), while CD8+CD16+natural killer (NK) lymphocytes and CD8+ T-cell receptor (TCR)-γ/δ+ T lymphocytes express an α/α homodimer (Mr 30 kDa). CD8+TCR-α/β+ T lymphocytes can express either an α/α homodimer or α/β heterodimer. The CD8 antigenic determinant binds to class I major histocompatibility (MHC) molecules, resulting in increased adhesion between the CD8+ T lymphocytes and target cells. Binding of the CD8 antigen to class I MHC molecules enhances the activation of resting T lymphocytes. The CD8 antigen is coupled to a protein tyrosine kinase, p56lck. The CD8:p56lck complex can play a role in T-lymphocyte activation through mediation of the interactions between the CD8 antigen and the CD3/TCR complex.

TRUSTED_SUSTAINABILITY

Specifications

Antígeno CD8
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon SK1
Concentración 50μg/mL
Conjugado APC-H7
Formulación Phosphate buffered saline with gelatin and 0.1% sodium azide.
Especie del huésped Mouse
Inmunógeno Human Peripheral Blood T Cells
Método de purificación Affinity Purified
Cantidad 100 Tests
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.