Learn More
CD71 Mouse anti-Human, Brilliant Violet 786, Clone: M-A712, BD
Descripción
The M-A712 monoclonal antibody specifically binds to CD71 which is also known as the transferrin receptor (TFR). This type II transmembrane glycoprotein is expressed on cells as a disulfide-linked homodimer comprised of 95 kDa monomers. CD71 is expressed on activated lymphocytes, monocytes, macrophages, erythroid progenitors, brain endothelium, and most proliferating cells. CD71 is not expressed on resting lymphocytes and is upregulated during lymphocyte responses to antigens or mitogens. Through an endocytic pathway, the transferrin receptor mediates cellular iron uptake by binding and internalizing iron that is bound to transferrin. After releasing iron within the low pH endosomal environment, transferrin and its receptor can be recycled to the cell surface.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon™ BV786 can be excited by the violet laser and detected in a filter used to detect Cy7™-like dyes (eg, 780/60-nm filter).
Especificaciones
Especificaciones
| Antígeno | CD71 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | M-A712 |
| Conjugado | Brilliant Violet 786 |
| Descripción | TFR; TFRC; Trfr; TfR1; Transferrin receptor protein 1 |
| Especie del huésped | Mouse |
| Inmunógeno | T-CLL and Jurkat Cells |
| Método de purificación | Affinity Purified |
| Cantidad | 50 Tests |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.