Learn More
anti-CD70, Human, Matched Antibody Pair, Abnova™
Código de producto. 16101874
Change view
Click to view available options
Cantidad:
1 Set
Tamaño de la unidad:
1 set
16101874 1 Set 1 set
1 options
Código de producto. 16101874
Marca: Abnova H00000970AP42.1Set
Includes
Capture antibody: mouse monoclonal anti-CD70, IgG1 Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-CD70, IgG1 Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
To detect and quantify protein level of human
Especificaciones
Antígeno CD70
Aplicaciones ELISA
Clasificación Monoclonal/Polyclonal
Conjugado Biotin
Descripción This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CD70.
génica CD70
Alias de gen CD27L/CD27LG/TNFSF7
Símbolos de los genes CD70
Especie del huésped Mouse
Cantidad 1 Set
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 970
Incluye Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CD70, IgG1 Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-CD70, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Especies diana Human
Contenido y almacenamiento Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Formulario Liquid
Mostrar más Mostrar menos
For Research Use Only