missing translation for 'onlineSavingsMsg'
Learn More

CD69 Mouse anti-Human, V450, Clone: FN50, BD

Código de producto. 15880449
Change view
Click to view available options
Cantidad:
50 Tests
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15880449 50 Tests cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15880449

Marca: BD Biosciences 560740

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The FN50 monoclonal antibody specifically binds to human CD69. CD69 is also known as activation-induced molecule (AIM), early activation antigen (EA-1), very early activation antigen (VEA), C-type lectin domain family 2 member C (CLEC2C), MLR-3, GP32/28 and Leu-23. CD69 is a transmembrane type II homodimer receptor. CD69 is comprised of disulfide-linked, differentially glycosylated core protein subunits that are approximately 28 and 34 kDa in size. Each subunit contains a C-type lectin domain. CD69 is expressed on activated T, B, and natural killer (NK) lymphocytes, thymocytes, neutrophils, eosinophils and platelets. In normal peripheral blood, a small and variable percentage of lymphocytes typically express detectable membrane CD69 antigen. Upon activation, CD69 antigen expression increases on lymphocytes. Peak CD69 expression generally occurs within 18 hours of activation, preceding the appearance of HLA-DR, IL-2Rα (CD25) and transferrin receptor (CD71). CD69 is highly expressed on the bright CD3+ subset of thymocytes. FN50 monoclonal antibody labels NK cells and most lymphocytes of the follicular mantle and perifollicular/interfollicular zone as well as germinal center T cells of lymph nodes and tonsils. Studies indicate that CD69 serves as a signaling receptor in the activation of a variety of cell types.

The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD69
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon FN50
Conjugado Violet Fluor 450 (V450)
Descripción AIM; CLEC2C; EA1; GP32/28; Leu23; MLR-3; VEA; BL-AC/P26
Formulación Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Especie del huésped Mouse
Método de purificación Affinity Purified
Cantidad 50 Tests
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.