Learn More
CD62E Mouse anti-Human, PE, Clone: 68-5H11, BD
Descripción
The 68-5H11 monoclonal antibody specifically binds to CD62E. This adhesion molecule is a 97-115 kDa type I transmembrane glycoprotein that is encoded by the SELE gene. CD62E is also known as E-selectin, Endothelial-leukocyte adhesion molecule-1 (ELAM-1), and Leukocyte endothelial cell adhesion molecule 2 (LECAM-2). CD62E is minimally expressed by unstimulated endothelium. Activated endothelial cells upregulate surface CD62E expression in response to various activators including inflammatory cytokines such as Interleukin-1 and Tumor Necrosis Factor. CD62E plays a role in leucocyte extravasation by promoting leucocyte rolling on activated endothelial cells during inflammation. CD62E may also play a role in the metastasis of certain tumor cells. The adhesion between endothelial CD62E molecules and a carbohydrate ligand on neutrophils is inhibited by the mAb 68-5H11.
Especificaciones
Especificaciones
| Antígeno | CD62E |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 68-5H11 |
| Conjugado | PE |
| Descripción | SELE; E-selectin; Selectin E; ELAM; ELAM1; ESEL; LECAM2 |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Método de purificación | Affinity Purified |
| Cantidad | 100 Tests |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.