missing translation for 'onlineSavingsMsg'
Learn More

CD58 (LFA-3) Mouse anti-Human, PE, Clone: L306.4, BD

Código de producto. 15835858
Change view
Click to view available options
Cantidad:
50 Tests
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15835858 50 Tests cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 15835858

Brand: BD Biosciences 340295

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD58 (Anti-LFA-3), clone L306.4, is derived from hybridization of mouse Sp2/0 myeloma cells with spleen cells from C3H/HeN mice immunized with Lc-LFA-3 mouse L-cell transfectant.

CD58 (Anti-LFA-3) recognizes a 40 to 65-kdalton cell-surface glycoprotein that is a member of the immunoglobulin superfamily. The CD58 antigen, known as the lymphocyte function–associated antigen-3 (LFA-3), mediates cellular adhesion and participates in signal transduction when it binds to its ligand, the CD2 antigen. Cellular interactions regulated by the CD58/CD2 antigens are involved in the antigen-independent adhesion pathway and cytotoxic T lymphocyte (CTL) activity. The CD58 antigen has two isoforms. One isoform is anchored in the cell membrane by a glycophosphatidyl inositol tail, while the other has a transmembrane hydrophobic segment and a cytoplasmic segment composed of 12 amino acids.

TRUSTED_SUSTAINABILITY

Specifications

Antígeno CD58
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon L306.4
Conjugado PE
Formulación Phosphate buffered saline with gelatin and 0.1% sodium azide.
Especie del huésped Mouse
Inmunógeno Lc-LFA-3 mouse L-cell transfectant
Cantidad 50 Tests
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.