Learn More
CD56 Mouse anti-Human, V450, Clone: B159, BD
Descripción
Reacts with the 220/135kDa heavily glycosylated antigen isoforms present on a subpopulation of peripheral blood large granular lymphocytes which demonstrate natural killer activity, but not on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neutral cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Host Species: Mouse
Clone: B159
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Especificaciones
Especificaciones
| Antígeno | CD56 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | B159 |
| Conjugado | Violet Fluor 450 (V450) |
| Descripción | NCAM1; NCAM-1; Neural cell adhesion molecule 1; NCAM; NKH1; MSK39 |
| Formulación | Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human NK Cells |
| Método de purificación | Affinity Purified |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.