missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD55 Matched Antibody Pair, Abnova™
Marca: Abnova H00001604-AP51.1Set
Includes: Capture antibody: mouse monoclonal anti-CD55 (100μg); Detection antibody: rabbit purified polyclonal anti-CD55 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Descripción
Host Species: Mouse, Rabbit
Cross Reactivity: Human
ELISA Pair
Especificaciones
| CD55 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CD55. | |
| CR/CROM/DAF/TC | |
| Mouse, Rabbit | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CD55 (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-CD55 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| CD55 | |
| CD55 | |
| 1 Set | |
| 1604 | |
| Human |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?