Learn More
CD49b (Pan-NK cells) Rat anti-Mouse, APC, Clone: DX5, BD
Descripción
The rat anti-mouse CD49b monoclonal antibody (clone DX5) specifically binds t o the integrin α2 chain (CD49b). C D49b is a 150 kDa transmembrane glycoprotein that non-covalently ass ociates with CD29 (integrin β1) to form the integrin α2β1 complex known as VLA-2. The rat anti-mouse CD49b antibody (clone DX5) has been reported to identify the majority of NK cells and a small T-cell subpopulation in most mouse strains (e.g., A/J, AKR, BALB/c, C3H/HeJ, C57BL/6, C57BL/10, C57BR, C58, CBA/Ca, DBA/1, DBA/2, SJL, SWR, 129/J, but not NOD). The DX5 antibody also recognizes platelets that express high levels of CD49b. Multiparameter flow cytometric analysis has demonstrated that most lymphocytes which express NK-1.1 (NKR-P1B and NKR-P1C), as detectable by mouse anti-mouse NK-1.1 antibody (clone PK136), also express the DX5 antigen. Small DX5+ NK-1.1- and DX5- NK-1.1+ cell subsets are found, especially among the CD3-positive cell population. Some CD49b+ NK cells have been reported to gradually lose reactivity with the rat anti-mouse CD49b antibody (clone DX5) when cultured in the presence of recombinant human IL-2. The resulting DX5-negative cells have weakened cytotoxic activity when compared to the remaining DX5+ cells. This indicates that the DX5 antibody distinguishes functional subsets of NK cells. No activation or blocking activity of the rat anti-mouse antibody (clone DX5) has been observed. Staining of splenic NK cells with this antibody reportedly can be blocked by hamster anti-mouse CD49b antibody (clone HMα2).
Especificaciones
Especificaciones
| Antígeno | CD49b (Pan NK) |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | DX5 |
| Concentración | 0.2mg/mL |
| Conjugado | APC |
| Descripción | itga2; Integrin alpha-2; DX5; Pan NK cell marker; VLAA2; VLA-2 alpha chain |
| Formulación | Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. |
| Especie del huésped | Rat |
| Inmunógeno | rIL-2n++agated NK1.1+ cells from C57BL/6 mice |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.