Learn More
CD45 Mouse anti-Human, FITC, Clone: HI30, BD
Código de producto. p-4327100
Change view
Click to view available options
Cantidad:
25 Tests
500 Tests
Tamaño de la unidad:
cada uno
15890649 25 Tests cada uno
15801419 500 Tests cada uno
2 options
Código de producto. 15890649
Marca: BD Biosciences 560976
Mouse Monoclonal Antibody
The HI30 monoclonal antibody specifically binds to the 180, 190, 205, 220 kDa protein isoforms of CD45. CD45 is encoded by the PTPRC (Protein tyrosine phosphatase receptor type C) gene. CD45, also known as the leukocyte common antigen (LCA), is a member of the protein tyrosine phosphatase (PTP) family. It is present on all human leukocytes including lymphocytes, monocytes, granulocytes, eosinophils, and thymocytes. CD45 is absent from circulating erythrocytes, platelets, or mature erythroid cells of bone marrow and non-hemopoietic tissues.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CD45
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon HI30
Conjugado FITC
Descripción PTPRC; CD45R; LCA; L-CA; Leukocyte Common Antigen; B220; T200; GP180; LY5
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Human Peripheral Blood Leucocytes
Método de purificación Affinity Purified
Cantidad 25 Tests
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Mostrar más Mostrar menos
For Research Use Only.