Learn More
CD41b Mouse anti-Human, Unlabeled, Clone: HIP2, BD
Descripción
The HIP2 monoclonal antibody specifically binds to CD41b. CD41b/CD41 is encoded by the ITGA2B gene and is also known as Platelet glycoprotein GPIIb and Integrin alpha IIb. CD41 is transmembrane glycoprotein that is post-translationally processed into a disulfide linked α-chain and β-chain. CD41 combines with CD61, otherwise known as Glycoprotein IIIa or β3 integrin, to form the CD41/CD61 (GPIIb/IIIa) complex. This complex is expressed on platelets and megakaryocytes and is involved in platelet activation, aggregation and binding to extracellular matrix proteins. Upon platelet activation, the CD41/CD61 complex can function as a receptor for fribrinogen, fibronectin and von Willebrand factor. In contrast to other CD41b antibodies, HIP2 does not detect anti-ZWa bound to GPIIb/IIIa. This antibody appears to be a weak aggregation inducer.
Especificaciones
Especificaciones
| Antígeno | CD41b |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | HIP2 |
| Concentración | 0.5mg/mL |
| Conjugado | Unconjugated |
| Descripción | ITGA2B; CD41; GP2B; GPIIb; GPalpha IIb; Platelet glycoprotein GPIIb |
| Formulación | Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Método de purificación | Affinity Purified |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.