missing translation for 'onlineSavingsMsg'
Learn More

CD4 Rat anti-Mouse, FITC, Clone: RM4-5, BD

Rat Monoclonal Antibody

101.50€ - 274.99€

Especificaciones

Antígeno CD4
Clon RM4-5
Concentración 0.5mg/mL
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15836958
Ver documentos
BD Biosciences
553046
0.1 mg
ONLINE EXCLUSIVE PRICE
102.00€
101.50€ / cada uno
Ahorro 0.50€
0.49% Descuento
Ver productos alternativos
Ahorre hasta 
»
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
10461654
Ver documentos
BD Biosciences
553047
0.5 mg
ONLINE EXCLUSIVE PRICE
275.00€
274.99€ / cada uno
Ahorro 0.01€
0.00% Descuento
Ver productos alternativos
Ahorre hasta 
»
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

The RM4-5 clone has been reported to react with the CD4 (L3T4) differentiation antigen expressed on most thymocytes, subpopulations of mature T lymphocytes (i.e., MHC class II-restricted T cells, including most T helper cells and immunosuppressive regulatory T cells), and a subset of NK-T cells. CD4 has also been reported to be detected on pluripotent hematopoietic stem cells, bone marrow myeloid and B-lymphocyte precursors, intrathymic lymphoid precursors, and a subset of splenic dendritic cells. CD4 has been reported to be expressed on the plasma membrane of mouse egg cells and is involved in adhesion of the egg to MHC class II-bearing sperm. CD4 is an antigen coreceptor on the T-cell surface which interacts with MHC class II molecules on antigen-presenting cells. It participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase lck. Purified RM4-5 mAb has been reported to block the binding of FITC-conjugated anti-mouse CD4 clones GK1.5 and H129.19, but not the RM4-4 clone. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: RM4-5
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse Thymocytes (BALB/c)

Flow Cytometry

Especificaciones

Especificaciones

CD4
0.5mg/mL
Monoclonal
Rat
Immunology
Aqueous buffered solution containing ≤0.09% sodium azide.
IgG2a κ
Affinity Purified
Cd4; CD4 antigen; L3T4; Ly-4; T-cell surface antigen T4/Leu-3
RM4-5
Flow Cytometry
FITC
RUO
Mouse
Mouse Thymocytes (BALB/c)
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documentos

Documentos

Product Certifications

For Research Use Only.

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado