Learn More
CD4 Mouse anti-Human, FITC, Clone: L200, BD
Descripción
The L200 monoclonal antibody specifically binds to the human form of the 56 kDa transmembrane glycoprotein, CD4, present on the T-helper/inducer subset of normal human donor peripheral blood lymphocytes. The L200 antibody also crossreacts with a subset of CD3-positive peripheral blood lymphocytes, but not monocytes, of both Rhesus and Cynomolgus Macaque monkeys. Crossreactivity on both lymphocytes and monocytes (weak) from Baboons is also observed. The distribution on lymphocytes is similar for both human and monkey cells, with the majority of CD4-positive lymphocytes being CD8-negative and lacking reactivity with antibodies to B- or NK-cell markers.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Especificaciones
Especificaciones
| Antígeno | CD4 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | L200 |
| Conjugado | FITC |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human HPB-ALL Cell Line |
| Método de purificación | Affinity Purified |
| Cantidad | 50 Tests |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.