Learn More
CD4 Rat anti-Mouse, APC-H7, Clone: GK1.5, BD
Descripción
The GK1.5 clone has been reported to react with the CD4 (L3T4) differentiation antigen expressed on most thymocytes, a subpopulation of mature T lymphocytes (i.e., MHC class II-restricted T cells, including most T helper cells), and a subset of NK-T cells. In addition, CD4 has also been reported to be detectable on pluripotent hematopoietic stem cells, bone marrow myeliod and B-lymphocyte precursors, intrathymic lymphoid precursors, and a subset of splenic dendritic cells. CD4 has also been reported to be expressed on the plasma membrane of mouse egg cells and is involved in adhesion of the egg to MHC class II-bearing sperm. CD4 is an antigen coreceptor on the T-cell surface which interacts with MHC class II molecules on antigen-presenting cells. It participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase lck. GK1.5 mAb reportedly blocks binding of the RM4-5 (Mfr. No. 553046/553047) and H129.19 (Mfr. No. 553650/553651), but not RM4-4 (Mfr. No. 553055) antibodies.
Host Species: Rat
Clone: GK1.5
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: Mouse CTL clone V4
Flow Cytometry, Intracellular Staining
Especificaciones
Especificaciones
| Antígeno | CD4 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | GK1.5 |
| Concentración | 0.2mg/mL |
| Conjugado | APC-H7 |
| Descripción | Cd4; CD4 antigen; L3T4; Ly-4; T-cell surface antigen T4/Leu-3 |
| Formulación | Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. |
| Especie del huésped | Rat |
| Inmunógeno | Mouse CTL clone V4 |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.