missing translation for 'onlineSavingsMsg'
Learn More

CD3ε Hamster anti-Mouse, PE, Clone: 145-2C11, BD

Código de producto. p-3969554
Change view
Click to view available options
Cantidad:
0.1 mg
0.2 mg
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15866968 0.1 mg cada uno
15876968 0.2 mg cada uno
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15866968

Marca: BD Biosciences 553063

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Hamster Monoclonal Antibody

The 145-2C11 antibody reacts with the 25-kDa ε chain of the T-cell receptor-associated CD3 complex, which is expressed on thymocytes, mature T lymphocytes, and NK-T cells. The cytoplasmic domain of CD3e participates in the signal transduction events which activate several cellular biochemical pathways as a result of antigen recognition. Soluble 145-2C11 antibody can activate either unprimed (naive) or primed (memory/preactivated) T cells in vivo or in vitro, in the presence of Fc receptor-bearing accessory cells. In contrast, plate-bound 145-2C11 can activate T cells in the absence of accessory cells. Soluble 145-2C11 antibody has been reported to induce re-directed lysis of Fc receptor-bearing target cells by CTL clones and can also block lysis of specific target cells by antigen-specific CTL's. Under some conditions, T-cell activation by 145-2C11 antibody has been reported to result in apoptotic cell death. The 145-2C11 antibody does not cross-react with rat leukocytes and it has been reported that pre-incubation of thymus cell suspensions at 37°C for 2-4 hours prior to staining enhances the ability of anti-CD3ε and anti-αβ TCR mAbs to detect the T-cell receptor on immature thymocytes. This antibody is routinely tested by flow cytometric analysis and is compatible with intracellular staining protocols using the BD Cytofix/Cytoperm™ Kit (Cat. No. 554714). Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Hamster
Clone: 145-2C11
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: H-2Kb specific cytotoxic T lymphocyte clone BM10-37

Flow Cytometry, Intracellular Staining

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD3 ε
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 145-2C11
Concentración 0.2mg/mL
Conjugado PE
Descripción CD3; CD3 epsilon; Cd3e; CD3ε; T3e
Formulación Aqueous buffered solution containing ≤0.09% sodium azide.
Especie del huésped Hamster
Inmunógeno H-2Kb specific cytotoxic T lymphocyte clone BM10-37
Método de purificación Affinity Purified
Cantidad 0.1 mg
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
Especies diana Mouse
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Mostrar más Mostrar menos

For Research Use Only.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.