Learn More
CD29 Rat anti-Mouse, Unlabeled, Clone: 9EG7, BD
Descripción
The 9EG7 antibody reacts with the 130kDa integrin β1 chain (CD29). CD29 is expressed on the cell surface as a heterodimer with one of the distinct integrin αchains. With α1 through α6 (CD49a through CD49f), it forms the VLA-1 through VLA-6 complexes, respectively, and with αv (CD51), it forms αvβ1 integrin. It also associates with the integrin α7 α8 and α9 chains in non-lymphoid tissues. As a result, CD29 has a broad tissue distribution, including lymphocytes, endothelia, smooth muscle, and epithelia. 9EG7 mAb has been shown to inhibit both the α6β1 mediated binding of lymphocytes to endothelial cells and the adhesion mediated by activated, but not unactivated, α4β1-integrin. The source of the immunogen was mouse lymph node-derived endothelial cell line TME.
Especificaciones
Especificaciones
| Antígeno | CD29 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 9EG7 |
| Concentración | 31.25μg/mL |
| Conjugado | Unconjugated |
| Descripción | Integrin β1 chain |
| Formulación | Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide. |
| Especie del huésped | Rat |
| Inmunógeno | Mouse endothelial cell line |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.