missing translation for 'onlineSavingsMsg'
Learn More

CD244 Mouse anti-Human, Unlabeled, Clone: 2-69, BD

Código de producto. 15836398
Change view
Click to view available options
Cantidad:
0.1 mg
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15836398 0.1 mg cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15836398 Proveedor BD Biosciences N.º de proveedor 550814

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The 2-69 monoclonal antibody specifically binds to 2B4 which is also known as CD244. CD244 is an approximately 63-70 kDa type I transmembrane glycoprotein. It is a SLAM submember (SLAMF4) and CD2 family member of the immunoglobulin superfamily. CD244 was originally identified in the mouse as a non-MHC-restricted cytotoxicity mediator present on NK cells and CD8+ T cells. In humans, CD244 is expressed on NK cells, γδ T cells, subsets of effector and memory CD8+ T cells, monocytes, eosinophils and basophils. This expression pattern suggests a broad role for CD244 in the regulation of leukocyte activation. CD244 binds CD48 with high affinity. CD244 may serve both stimulatory and inhibitory functions depending on a number of factors. These include the intensity and duration of CD244 interactions with its ligands as well as the presence of other signals generated through different receptor-ligand interactions. 2B4 was clustered as CD244 in the VIIth HLDA workshop.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD244
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon Feb-69
Concentración 0.5mg/mL
Conjugado Unconjugated
Descripción 2B4; SLAMF4; NAIL; NK cell activation inducing ligand ; NKR2B4; Nmrk
Formulación Aqueous buffered solution containing ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Human NK Cells
Método de purificación Affinity Purified
Cantidad 0.1 mg
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C
Isotype IgG2a κ
Mostrar más Mostrar menos

For Research Use Only.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.