missing translation for 'onlineSavingsMsg'
Learn More

CD19 Rat anti-Mouse, PE, Clone: 1D3, BD

Código de producto. p-3844152
Change view
Click to view available options
Cantidad:
0.1 mg
25 μg
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15811289 25 μg cada uno
15848908 0.1 mg cada uno
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15811289

Marca: BD Biosciences 561736

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

The 1D3 antibody reacts with CD19, a B lymphocyte-lineage differentiation antigen. CD19, a 95kDa transmembrance glycoprotein, is a member of the immunoglobulin superfamily and is expressed throughout B-lymphocyte development from the pro-B cell through the mature B-cell stages. Terminally differentiated plasma cells do not express CD19. On the surface of mature B cells, the CD19 molecule associates with CD21 (CR-2) and CD81 (TAPA-1), and this multimolecular complex synergizes with surface immunoglobulin to promote cellular activation. Studies with CD19-deficient mice have suggested that the level of CD19 expression affects the generation and maturation of B cells in the bone marrow and periphery. B-1 lineage B cells, also known as CD5+ B cells, are drastically reduced or absent in CD19-deficient mice. Increased levels of CD19 expression correlate with increased frequencies of peritonal and splenic B-1 cells and reduced numbers of conventional B lymphocytes in the periphery. CD19 participates in B-lymphocyte development, B-cell activation, maturation of memory B cells and regulation of tolerance. CD19 has also been detected on peritoneal mast cells, co-localized with CD21/CD35, and it is proposed to play a role in complement-mediated mast-cell activation. This antibody is routinely tested by flow cytometric analysis and is compatible with intracellular staining protocols using the BD Cytofix/Cytoperm™ Kit (Mfr. No. 554714). Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: 1D3
Isotype: IgG2a, κ
Cross Reactivity: Mouse

Flow Cytometry

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD19
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 1D3
Concentración 0.2mg/mL
Conjugado PE
Formulación Aqueous buffered solution containing ≤0.09% sodium azide.
Especie del huésped Rat
Inmunógeno Mouse CD19 Transfected Cell Line
Método de purificación Affinity Purified
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Mouse
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Mostrar más Mostrar menos

For Research Use Only.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.