Learn More
CD178 Mouse anti-Human, Unlabeled, Clone: NOK-2, BD
Descripción
Fas (APO-1, CD95) is a 45kDa cell surface protein that mediates apoptosis when cross-linked with agonistic anti-Fas antibodies or Fas ligand (FasL). Fas belongs to the TNF (tumor necrosis factor)/NGF (nerve growth factor) receptor family, and is expressed in various tissue and cells including the thymus, liver, ovary, and lung. FasL is a 40kDa TNF family membrane protein that induces apoptosis by binding to Fas, its cell-surface receptor. FasL is expressed on activated T and NK cells. Both Fas and FasL are thought to play an important role in the apoptotic processes that take place during T cell development. NOK-2 recognizes human FasL. It recognizes both the membrane bound (FasL) and soluble (sFasL) forms. L5178Y mouse T lymphoma cells expressing recombinant human FasL were used as immunogen. FasL and sFasL migrate at reduced molecular weights of 40 and 26kDa, respectively. However, the molecular weights observed in a particular sample may vary according to FasL and sFasL glycosylation and breakdown patterns as described in Tanaka et al.
Especificaciones
Especificaciones
| Antígeno | CD178 |
| Aplicaciones | Immunoprecipitation |
| Clasificación | Monoclonal |
| Clon | NOK-2 |
| Concentración | 0.5mg/mL |
| Conjugado | Unconjugated |
| Descripción | Fas Ligand, CD95 Ligand |
| Formulación | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | L51788Y Mouse T Lymphoma Cells Expressing Recombinant Human FasL |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.