Learn More
CD15 Mouse anti-Human, FITC, Clone: HI98 (also known as HIM1), BD
Descripción
Reacts with 3-fucosyl-N-acetyllactosamine (3-FAL), a 220kDa carbohydrate structure, also called X-hapten, SSEA-1, CD15 or Lewis X. 3-FAL is expressed on >95% of granulocytes, including neutrophils and eosinophils, and to a varying degree on monocytes, but not on lymphocytes or basophils. CD15 plays a role in mediating phagocytosis, bactericidal activity and chemotaxis. This antibody is also suitable for staining formalin-fixed, paraffin-embedded tissue sections without pretreatment. Since the Abs are recognizing a carbohydrate epitope (3-fucosyl-N-acetyllactosamine) they also should work across species and not only for human.
Flow Cytometry
Especificaciones
Especificaciones
| Antígeno | CD15 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | HI98 (also known as HIM1) |
| Conjugado | FITC |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human Mononuclear Cells |
| Método de purificación | Affinity Purified |
| Cantidad | 25 Tests |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.