Learn More
CD15 Mouse anti-Human, Unlabeled, Clone: MMA, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 347420
Descripción
CD15 recognizes a human myelomonocytic antigen. The structure recognized by CD15 antibodies is lacto-N-fucopentose III.The CD15 antigen is present on greater than 95% of mature peripheral blood eosinophils and neutrophils and is present at low density on circulating monocytes.In lymphoid tissue, CD15 reacts with Reed-Sternberg cells of Hodgkin's disease and with granulocytes. However, CD15 reacts with few tissue macrophages and does not react with dendritic cells.
Especificaciones
| CD15 | |
| Monoclonal | |
| 50μg/mL | |
| HEPES buffer with gelatin and 0.1% sodium azide. | |
| U-937 histiocytic cell line | |
| 100 Tests | |
| Cell Biology | |
| Human | |
| IgM κ |
| Flow Cytometry | |
| MMA | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
For Research Use Only.