missing translation for 'onlineSavingsMsg'
Learn More

VE-cadherin/CD144 Polyclonal Antibody, Invitrogen™

Código de producto. 10345114
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
10345114 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 10345114

Marca: Invitrogen PA184328

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

VE-cadherin/CD144 Antibody, PA1-84328, from Invitrogen™. Species Reactivity: Bovine, Human, Mouse, Rat; Applications: ELISA, FACS, Functional Assay, IHC (Frozen), WB.

VE-cadherin/CD144 Antibody, PA1-84328, from Invitrogen. Species Reactivity: Bovine, Human, Mouse, Rat; Applications: ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Western Blot.

This gene is a classical cadherin from the cadherin superfamily and is located in a six-cadherin cluster in a region on the long arm of chromosome 16 that is involved in loss of heterozygosity events in breast and prostate cancer. The encoded protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Functioning as a classic cadherin by imparting to cells the ability to adhere in a homophilic manner, the protein may play an important role in endothelial cell biology through control of the cohesion and organization of the intercellular junctions. An alternative splice variant has been described but its full length sequence has not been determined.
Host Species: Rabbit
Cross Reactivity: Bovine, Human, Mouse, Rat
Isotype: IgG
Immunogen: Recombinant protein fragments corresponding to amio acids 1-258 of VE-cadherin.
Formulation: 100μg purified antibody (1mg/mL) in PBS and no preservative
Synonyms: Cadherin-5, Vascular endothelial cadherin, VE-cadherin, 7B4 antigen, CD144 antigen, CDH5
Uniprot Accession: P33151, P55284, Q6URK6
Entrez GeneID: 1003, 12562, 307618, 414735
Gene Symbol: CDH5
This antibody does not cross react with N-cadherin, which is also expressed by endothelial cells. It has also been reported to block adhesive properties of VE-cadherin. For FACS analysis, use 10ul of the suggested working dilution to label 1x10^6 cells in 100ul.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno VE-cadherin/CD144
Aplicaciones ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
N.º de referencia del gen P33151, P55284, Q6URK6
Alias de gen Cadherin-5, Vascular endothelial cadherin, VE-cadherin, 7B4 antigen, CD144 antigen, CDH5
Símbolos de los genes CDH5
Especie del huésped Rabbit
Inmunógeno Recombinant protein fragments corresponding to amio acids 1-258 of VE-cadherin.
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1003, 12562, 307618, 414735
Especies diana Bovine, Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C, Avoid Freeze/Thaw Cycles
Formulario Purified
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.