Learn More
CD140b (pY857) Mouse, PE, Clone: J24-618, BD
Descripción
Platelet-derived growth factor (PDGF) is a potent mitogen for cells of mesenchymal origin and exerts its effects by binding to the PDGF receptor (PDGFR), a transmembrane protein tyrosine kinase. PDGFR is composed of PDGFRα (CD140a) and/or PDGFRβ (CD140b) polypeptides. Both PDGF and PDGFR consist of subunits that form homo- or heterodimers with varying specificities: PDGF-AA binds only to αα PDGFR, PDGF-AB binds to both αα and αβ PDGFR, and PDGF-BB binds to all three PDGFRs. Ligand binding induces dimerization and activation of the receptor. Upon activation, CD140b is phosphorylated at multiple tyrosine sites and, in turn, an intracellular phosphorylation cascade is initiated. PDGFR localizes primarily to membrane invaginations termed caveolae, compartments that are enriched in several of its downstream effectors, including phosphatidylinositol 3'-kinase, Src, and phospholipase C-γ.
The J24-618 monoclonal antibody recognizes the phosphorylated tyrosine 857 (pY857) in the tyrosine kinase domain 2 of CD140b, which is required for maximal receptor kinase activity. The orthologous phosphorylation site in mouse PDGFRβ is Y856.
Especificaciones
Especificaciones
| Antígeno | CD140b (pY857) |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | J24-618 |
| Conjugado | PE |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Phosphorylated Human PDGFRβ Peptide |
| Método de purificación | Affinity Purified |
| Cantidad | 50 Tests |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.