Learn More
CD132 Mouse anti-Human, Brilliant Violet 421, Clone: AG184, BD
Descripción
TheAG184 monoclonal antibody specifically binds to the 65-70 kDa common γ subunit (γc) that is shared by the IL-2, IL-4, IL-7, IL-9, IL-15 and IL-21 receptor complexes. The γc receptor is a type 1 transmembrane glycoprotein that is constitutively expressed by most peripheral T and B lymphocytes, NK cells, monocytes and granulocytes. The cytoplasmic domain of the γc chain plays an important role in cytokine-mediated signal transduction. By immunofluorescent staining and flow cytometric analyses, the AG184 antibody has been shown to specifically recognize human γc expressed by cell lines including human γc gene-transfected cell lines which are known to express the human γc chain. The AG184 antibody can bind the γc chain in the receptors complexed with IL-2, IL-4, or IL-7, indicating that the antibody recognizes an epitope which is distinct from the cytokine binding site of the γc chain.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon™ BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon™ BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.
Especificaciones
Especificaciones
| Antígeno | CD132 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | AG184 |
| Conjugado | Brilliant Violet 421 |
| Descripción | IL2RG; IL-2RG; IL-2Rγ; Common gamma chain; γc; CIDX; SCIDX; SCIDX1; IMD4 |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Método de purificación | Affinity Purified |
| Cantidad | 50 Tests |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.