Learn More
CD123 Mouse anti-Human, PE-Cy5, Clone: 9F5, BD
Código de producto. 15890679
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
cada uno
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
15890679 25 Tests cada uno
15896498 100 Tests cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 15890679 Proveedor BD Biosciences N.º de proveedor 561009
Mouse Monoclonal Antibody
The 9F5 monoclonal antibody specifically binds to CD123. CD123 is the 70 kDa IL-3 receptor α; chain (IL-3Rα;) that associates with the 120-140 kDa β; subunit (CD131/Common β;-chain/β;c) to form the functional IL-3 receptor complex. The β;c chain is also shared with distinct α; chain subunits to form the functional heterodimeric receptors for interleukins IL-5 and GM-CSF. IL-3Rα; is expressed on a subset of peripheral blood dendritic cells, myeloid precursors, basophils, mast cells, macrophages, and megakaryocytes. Reports indicate that IL-3Rα; is also expressed on lymphocytes. The IL-3R plays an important role in hematopoietic progenitor cell growth and differentiation. This antibody does not block binding of IL-3 to the IL-3 receptor.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CD123
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 9F5
Conjugado PE-Cyanine5
Descripción IL-3 Receptor α chain
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Human IL-3Rα Transfected Cell Line
Método de purificación Affinity Purified
Cantidad 25 Tests
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Mostrar más Mostrar menos
For Research Use Only.