Learn More
CD107a Mouse anti-Human, FITC, Clone: H4A3, BD
Descripción
Reacts with the heavily glycosylated 110kDa lysosomal-associated membrane protein, LAMP-1. LAMP-1 is a widely expressed intracellular antigen. It is also expressed on the surface of activated platelets, PHA-activated lymphocytes and some tumor cell lines, including U937 and KG1a. LAMP-1 has been shown to be a ligand for E-selectin-mediated cell adhesion. LAMP-1 and LAMP-2 (CD107b) are carriers for poly-N-acetyllactosamines, and are able to display sialyl Le[x] termini. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: H4A3
Isotype: IgG1 κ
Species Reactivity: Human
Intracellular Staining
Especificaciones
Especificaciones
| Antígeno | CD107a |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | H4A3 |
| Conjugado | FITC |
| Descripción | LAMP1; LAMP-1; LAMPA; LGP120 |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human Adult Adherent Peripheral Blood Cells |
| Método de purificación | Affinity Purified |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.